missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Methyl Sulfide 98.0+%, TCI America™

Catalog No. p-7119072
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14755ML 5 mL
B147525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14755ML Supplier TCI America Supplier No. B14755ML
Only null left

Chemical Identifiers

CAS 6163-64-0
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00008840
InChI Key CJFVCTVYZFTORU-UHFFFAOYSA-N
Synonym tert-butyl methyl sulfide, propane, 2-methyl-2-methylthio, sulfide, tert-butyl methyl, methyl tert-butyl sulfide, 3,3-dimethyl-2-thiabutane, 2-methyl-2-methylsulfanyl propane, t-butyl methyl sulfide, methyl t-butyl sulfide, tert-butyl methyl sulfane, acmc-1b7tx
PubChem CID 138679
IUPAC Name 2-methyl-2-(methylsulfanyl)propane
SMILES CSC(C)(C)C

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 mL
UN Number 1993
Formula Weight 104.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Methyl Sulfide