Learn More
tert-Butyl methyl ether, ACS reagent
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Supplier: Thermo Scientific Chemicals 447470025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| tert-Butyl methyl ether | |
| 1634-04-4 | |
| 54.0°C to 56.0°C | |
| -28°C | |
| 1.3675 to 1.3695 | |
| C5H12O | |
| 0.36 mPa.s (20°C) | |
| Solubility aq. soln.: 51 g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5 g/100 g | |
| COC(C)(C)C | |
| 88.15 | |
| ACS Reagent |
| -110.0°C | |
| 0.7404 g/mL | |
| 2.5 L | |
| Glass Bottle | |
| 0.7404 | |
| (CH3)3COCH3 | |
| 15, 6106 | |
| BZLVMXJERCGZMT-UHFFFAOYSA-N | |
| 2-methoxy-2-methylpropane | |
| 88.15 |
Chemical Identifiers
| 1634-04-4 | |
| 88.15 | |
| 2-methoxy-2-methylpropane |
| C5H12O | |
| BZLVMXJERCGZMT-UHFFFAOYSA-N | |
| COC(C)(C)C |
Safety and Handling
GHS H Statement Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.
No smoking.
Ground and bond container and receiving equipment
GHS Signal Word: Danger
EINECSNumber : 216-653-1
RUO – Research Use Only