missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, ACS reagent

Catalog No. p-7214372
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447470010 1 L
AC447470025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447470010 Supplier Thermo Scientific Chemicals Supplier No. 447470010
Only null left

CAS: 1634-04-4 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
IUPAC Name 2-methoxy-2-methylpropane
SMILES COC(C)(C)C

Specifications

Melting Point -110.0°C
Density 0.7404 g/mL
Boiling Point 54.0°C to 56.0°C
Quantity 1 L
Flash Point -28°C
Packaging Glass Bottle
Refractive Index 1.3675 to 1.3695
Specific Gravity 0.7404
Linear Formula (CH3)3COCH3
Viscosity 0.36 mPa.s (20°C)
Merck Index 15, 6106
Solubility Information Solubility aq. soln.: 51 g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5 g/100 g
Formula Weight 88.15
Grade ACS Reagent
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes skin irritation.

Highly flammable liquid and vapor.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

Ground and bond container and receiving equipment
EINECSNumber 216-653-1

RUO – Research Use Only