missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl isothiocyanate, 97%
Description
tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store under dry inert gas. Incompatible with amines, alcohols and water.
tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store under dry inert gas. Incompatible with amines, alcohols and water.
Specifications
Specifications
| Melting Point | 10°C to 12°C |
| Density | 0.908 |
| Boiling Point | 141°C to 145°C |
| Flash Point | 38°C (100°F) |
| Odor | Pungent |
| Refractive Index | 1.481 |
| Quantity | 5 g |
| UN Number | UN2929 |
| Beilstein | 1361854 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H226-H301+H311+H331-H314 |
| Hazard Statement | GHS H Statement H300-H310-H330-H314-H318-H226 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S. |
| EINECSNumber | 209-682-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only