missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl isothiocyanate, 97%

Catalog No. AAL0266806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0266806 5 g
AAL0266814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0266806 Supplier Thermo Scientific Chemicals Supplier No. L0266806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 590-42-1 | C5H9NS | 115.19 g/mol

tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Incompatible with amines, alcohols and water.

Chemical Identifiers

CAS 590-42-1
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00004816
InChI Key ZFWFRTVIIMTOLY-UHFFFAOYSA-N
Synonym tert-butyl isothiocyanate, t-butyl isothiocyanate, propane, 2-isothiocyanato-2-methyl, tert-butylisothiocyanate, tert.-butyl isothiocyanate, 2-isothiocyanato-2-methyl-propane, tert butyl isothiocyanate, tert-butyliso thiocyanate, butyl isothiocyanate, tert, t-butylisothiocyanate
PubChem CID 11544
IUPAC Name 2-isothiocyanato-2-methylpropane
SMILES CC(C)(C)N=C=S

Specifications

Melting Point 10°C to 12°C
Density 0.908
Boiling Point 141°C to 145°C
Flash Point 38°C (100°F)
Odor Pungent
Refractive Index 1.481
Quantity 5 g
UN Number UN2929
Beilstein 1361854
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 115.2
Percent Purity 97%
Chemical Name or Material tert-Butyl isothiocyanate
Show More Show Less
Hazard Category H226-H301+H311+H331-H314
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 209-682-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only