missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydrogen phthalate, 97%

Catalog No. p-7044865
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0853406 5 g
AAL0853414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0853406 Supplier Thermo Scientific Chemicals Supplier No. L0853406

CAS: 33693-84-4 | C12H14O4 | 222.24 g/mol

It finds its application in the resolution and reactions of tertiary alcohols : 2-phenylbutan-2-ol and 3-methylhexan-3-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the resolution and reactions of tertiary alcohols : 2-phenylbutan-2-ol and 3-methylhexan-3-ol.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 33693-84-4
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00043711
InChI Key PBUQZKXKYSAJDO-UHFFFAOYSA-N
Synonym tert-butyl hydrogen phthalate, 2-tert-butoxycarbonyl benzoic acid, unii-8390xkx0sq, 2-tert-butyl oxycarbonyl benzoic acid, t-monobutyl phthalate, t-butyl hydrogen phthalate, acmc-1ad5b, dsstox_cid_27150, dsstox_rid_82154, dsstox_gsid_47150
PubChem CID 134976
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonyl]benzoic acid
SMILES CC(C)(C)OC(=O)C1=CC=CC=C1C(=O)O

Specifications

Melting Point 84°C to 88°C
Quantity 5 g
Beilstein 2263386
Solubility Information Insoluble in water.
Formula Weight 222.24
Percent Purity 97%
Chemical Name or Material tert-Butyl hydrogen phthalate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only