missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Glycidyl Ether 96.0+%, TCI America™

Catalog No. B184025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B184025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B184025ML Supplier TCI America Supplier No. B184025ML
Only null left

Chemical Identifiers

CAS 7665-72-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00010050
InChI Key SFJRUJUEMVAZLM-UHFFFAOYNA-N
Synonym 1-tert-Butoxy-2,3-epoxypropane
PubChem CID 24314
IUPAC Name 2-[(tert-butoxy)methyl]oxirane
SMILES CC(C)(C)OCC1CO1

Specifications

Color Colorless
Boiling Point 152°C
Quantity 25 mL
UN Number 1993
Formula Weight 130.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Glycidyl Ether
RTECSNumber RR0475000
TSCA Yes