CAS RN 7665-72-7
CAS RN 7665-72-7
IUPAC Name:
2-[(tert-butoxy)methyl]oxirane
Synonyms:
2-(tert-butoxymethyl)oxirane
Molecular Weight (g/mol):
130.19
Molecular Formula:
C7H14O2
InChi Key:
SFJRUJUEMVAZLM-UHFFFAOYNA-N
SMILES:
CC(C)(C)OCC1CO1
1
–
1
of
1
results
1
tert-Butyl Glycidyl Ether 96.0+%, TCI America™
CAS: 7665-72-7 Molecular Formula: C7H14O2 Molecular Weight (g/mol): 130.19 MDL Number: MFCD00010050 InChI Key: SFJRUJUEMVAZLM-UHFFFAOYNA-N Synonym: 1-tert-Butoxy-2,3-epoxypropane PubChem CID: 24314 IUPAC Name: 2-[(tert-butoxy)methyl]oxirane SMILES: CC(C)(C)OCC1CO1