missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Chloroacetate 97.0+%, TCI America™

Catalog No. p-7124884
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C126725ML 25 mL
C1267500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C126725ML Supplier TCI America Supplier No. C126725ML
Only null left

Chemical Identifiers

CAS 107-59-5
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00000930
InChI Key KUYMVWXKHQSIAS-UHFFFAOYSA-N
Synonym tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate
PubChem CID 66052
IUPAC Name tert-butyl 2-chloroacetate
SMILES CC(C)(C)OC(=O)CCl

Specifications

Color Colorless
Boiling Point 60°C
Quantity 25 mL
UN Number 3272
Formula Weight 150.60
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Chloroacetate
EINECSNumber (2)-1149
RTECSNumber AF8985500
TSCA Yes