accessibility menu, dialog, popup

UserName

CAS RN 107-59-5

IUPAC Name: tert-butyl 2-chloroacetate
Synonyms: tert-butyl 2-chloroacetate
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: KUYMVWXKHQSIAS-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CCl
Boiling Point 
  • (3)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)