missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl chloroacetate, 98%

Catalog No. p-7022871
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1583314 25 g
AAA1583322 100 g
AAA1583336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1583314 Supplier Thermo Scientific Chemicals Supplier No. A1583314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-59-5 | C6H11ClO2 | 150.602 g/mol

tert-Butyl chloroacetate is used in the synthesis of imidazol-1-yl-acetic acid hydrochloride, cis-disubstituted aziridine ester via aza-Darzens reaction and 1,10-diaza-18-crown-6 based sensors bearing a coumarin fluorophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl chloroacetate is used in the synthesis of imidazol-1-yl-acetic acid hydrochloride, cis-disubstituted aziridine ester via aza-Darzens reaction and 1,10-diaza-18-crown-6 based sensors bearing a coumarin fluorophore.

Solubility
Decomposes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 107-59-5
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00000930
InChI Key KUYMVWXKHQSIAS-UHFFFAOYSA-N
Synonym tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate
PubChem CID 66052
IUPAC Name tert-butyl 2-chloroacetate
SMILES CC(C)(C)OC(=O)CCl

Specifications

Density 1.053
Boiling Point 155°C
Flash Point 46°C (115°F)
Odor Ester-like
Refractive Index 1.423
Linear Formula ClCH2CO2C(CH3)3
Quantity 25 g
UN Number UN2929
Beilstein 1753006
Merck Index 14,1563
Solubility Information Decomposes in water.
Formula Weight 150.61
Percent Purity 98%
Chemical Name or Material tert-Butyl chloroacetate
Show More Show Less
Hazard Category H226-H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 203-506-1
RTECSNumber AF8985500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only