Learn More
tert-Butyl bromide, 98+%, stab. with potassium carbonate
CAS: 507-19-7 | C4H9Br | 137.02 g/mol
$125.86 - $416.62
Chemical Identifiers
| CAS | 507-19-7 |
|---|---|
| Molecular Formula | C4H9Br |
| Molecular Weight (g/mol) | 137.02 |
| MDL Number | MFCD00000125 |
| InChI Key | RKSOPLXZQNSWAS-UHFFFAOYSA-N |
| Synonym | tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane |
| PubChem CID | 10485 |
| IUPAC Name | 2-bromo-2-methylpropane |
| SMILES | CC(C)(C)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1372722
|
Thermo Scientific Chemicals
A1372722 |
100 g |
Each for $125.86
|
|
|||||
|
AAA1372736
|
Thermo Scientific Chemicals
A1372736 |
500 g |
Each for $416.62
|
|
|||||
Description
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.
Solubility
Immiscible with water.
Notes
Incompatible with strong bases and strong oxidizing agents.
Chemical Identifiers
| 507-19-7 | |
| 137.02 | |
| RKSOPLXZQNSWAS-UHFFFAOYSA-N | |
| 10485 | |
| CC(C)(C)Br |
| C4H9Br | |
| MFCD00000125 | |
| tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane | |
| 2-bromo-2-methylpropane |
Specifications
| 507-19-7 | |
| 1.210 | |
| 18°C (64°F) | |
| C4H9Br | |
| MFCD00000125 | |
| UN2342 | |
| 14,1555 | |
| Immiscible with water. | |
| CC(C)(C)Br | |
| 137.02 | |
| 137.02 | |
| Liquid |
| °C to 20°C | |
| 74°C to 76°C | |
| 98%, stabilized with potassium carbonate | |
| 1.428 | |
| 100 g | |
| 1730892 | |
| tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane | |
| RKSOPLXZQNSWAS-UHFFFAOYSA-N | |
| 2-bromo-2-methylpropane | |
| 10485 | |
| ≥98% | |
| tert-Butyl Bromide, stabilized with potassium carbonate |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BROMOMETHYLPROPANES
EINECSNumber : 208-065-9
RTECSNumber : TX4150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only