missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl bromide, 98+%, stab. with potassium carbonate

Catalog No. AAA1372736
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1372736 500 g
AAA1372722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1372736 Supplier Thermo Scientific Chemicals Supplier No. A1372736
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 507-19-7 | C4H9Br | 137.02 g/mol

tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.

Solubility
Immiscible with water.

Notes
Incompatible with strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 507-19-7
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000125
InChI Key RKSOPLXZQNSWAS-UHFFFAOYSA-N
Synonym tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane
PubChem CID 10485
IUPAC Name 2-bromo-2-methylpropane
SMILES CC(C)(C)Br

Specifications

Melting Point -20°C
Density 1.21
Boiling Point 74°C to 76°C
Flash Point 18°C (64°F)
Assay Percent Range 98%, stabilized with potassium carbonate
Refractive Index 1.428
Quantity 500 g
UN Number UN2342
Beilstein 1730892
Merck Index 14,1555
Solubility Information Immiscible with water.
Formula Weight 137.02
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material tert-Butyl bromide, stabilized with potassium carbonate
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BROMOMETHYLPROPANES
EINECSNumber 208-065-9
RTECSNumber TX4150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only