missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl [Bis(4-methoxyphenyl)phosphinyloxy]carbamate 95.0+%, TCI America™

Catalog No. p-7119809
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28571G 1 g
B28575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28571G Supplier TCI America Supplier No. B28571G
Only null left

Chemical Identifiers

CAS 619333-95-8
Molecular Formula C19H24NO6P
Molecular Weight (g/mol) 393.38
MDL Number MFCD08276321
InChI Key BNMBQDIFMJFRAF-UHFFFAOYSA-N
Synonym [Bis(4-methoxyphenyl)phosphinyloxy]carbamic Acid tert-Butyl Ester, O-[Bis(4-methoxyphenyl)phosphinyl]-N-Boc-hydroxylamine, O-[Bis(4-methoxyphenyl)phosphinyl]-N-(tert-butoxycarbonyl)hydroxylamine
PubChem CID 44629804
IUPAC Name tert-butyl N-{[bis(4-methoxyphenyl)phosphoroso]oxy}carbamate
SMILES COC1=CC=C(C=C1)P(=O)(ONC(=O)OC(C)(C)C)C1=CC=C(OC)C=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 393.38
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl [Bis(4-methoxyphenyl)phosphinyloxy]carbamate
TSCA No
Recommended Storage Refrigerator