missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Benzoate 98.0+%, TCI America™

Catalog No. p-7118331
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B006725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B006725ML Supplier TCI America Supplier No. B006725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 774-65-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00048244
InChI Key LYDRKKWPKKEMNZ-UHFFFAOYSA-N
Synonym Benzoic Acid tert-Butyl Ester
PubChem CID 69886
ChEBI CHEBI:34034
IUPAC Name tert-butyl benzoate
SMILES CC(C)(C)OC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 54°C
Quantity 25 mL
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Benzoate
EINECSNumber (3)-1356
TSCA No