missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Alcohol 99.0+%, TCI America™

Supplier:  TCI America B0706500ML

 View more versions of this product

Catalog No. B0706500ML

Only null left

Description

Description

Specifications

Specifications

tert-Butyl Alcohol
75-65-0
C4H10O
1120
DKGAVHZHDRPRBM-UHFFFAOYSA-N
2-methylpropan-2-ol
6386
74.12
500 mL
White
82°C
MFCD00004464
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CC(C)(C)O
74.123
CHEBI:45895
≥99.0% (GC)
Crystal-Lump at 20°C

Chemical Identifiers

75-65-0
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
6386
2-methylpropan-2-ol
C4H10O
MFCD00004464
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CHEBI:45895
CC(C)(C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3049

RTECSNumber : EO1925000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions