missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Alcohol 99.0+%, TCI America™

Catalog No. B070625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B070625ML 25 mL
B0706500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B070625ML Supplier TCI America Supplier No. B070625ML

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Color White
Boiling Point 82°C
UN Number 1120
Formula Weight 74.12
Percent Purity ≥99.0% (GC)
Quantity 25 mL
Physical Form Crystal-Lump at 20°C
Chemical Name or Material tert-Butyl Alcohol
EINECSNumber (2)-3049
RTECSNumber EO1925000
TSCA Yes