missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Acetoacetate 95.0+%, TCI America™
$42.36 - $200.21
Chemical Identifiers
| CAS | 1694-31-1 |
|---|---|
| Molecular Formula | C8H14O3 |
| Molecular Weight (g/mol) | 158.197 |
| MDL Number | MFCD00008811 |
| InChI Key | JKUYRAMKJLMYLO-UHFFFAOYSA-N |
| Synonym | tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate |
| PubChem CID | 15538 |
| IUPAC Name | tert-butyl 3-oxobutanoate |
| SMILES | CC(=O)CC(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A081625ML
|
TCI America
A081625ML |
25 mL |
Each for $42.36
|
|
|||||
|
A0816100ML
|
TCI America
A0816100ML |
100 mL |
Each for $78.93
|
|
|||||
|
A0816500ML
|
TCI America
A0816500ML |
500 mL |
Each for $200.21
|
|
|||||
Chemical Identifiers
| 1694-31-1 | |
| 158.197 | |
| JKUYRAMKJLMYLO-UHFFFAOYSA-N | |
| 15538 | |
| CC(=O)CC(=O)OC(C)(C)C |
| C8H14O3 | |
| MFCD00008811 | |
| tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate | |
| tert-butyl 3-oxobutanoate |
Specifications
| 1694-31-1 | |
| 184°C | |
| MFCD00008811 | |
| tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate | |
| CC(=O)CC(=O)OC(C)(C)C | |
| 158.197 | |
| 158.20 | |
| Liquid |
| Yellow | |
| C8H14O3 | |
| 25 mL | |
| JKUYRAMKJLMYLO-UHFFFAOYSA-N | |
| tert-butyl 3-oxobutanoate | |
| 15538 | |
| ≥95.0% (GC) | |
| tert-Butyl Acetoacetate |
Safety and Handling
EINECSNumber : (2)-1475&(2)-1505
TSCA : Yes