accessibility menu, dialog, popup

UserName

CAS RN 1694-31-1

IUPAC Name: tert-butyl 3-oxobutanoate
Synonyms: tert-butyl 3-oxobutanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: JKUYRAMKJLMYLO-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)OC(C)(C)C
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)