missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl[4-(dimethoxymethyl)phenoxy]dimethylsilane 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 5 g |
| Formula Weight | 282.46 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | tert-Butyl[4-(dimethoxymethyl)phenoxy]dimethylsilane |
Safety and Handling
| TSCA | No |