missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, 97%

Catalog No. p-7035877
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5410003 1 g
AAH5410006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5410003 Supplier Thermo Scientific Chemicals Supplier No. H5410003
Only null left

CAS: 114772-40-6 | C18H19BrO2 | 347.252 g/mol

1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.

Solubility
Soluble in chloroform, ethyl acetate, and DMSO.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 114772-40-6
Molecular Formula C18H19BrO2
Molecular Weight (g/mol) 347.252
MDL Number MFCD06657561
InChI Key YHXCWNQNVMAENQ-UHFFFAOYSA-N
Synonym 2-boc-4'-bromomethyl biphenyl, tert-butyl 4'-bromomethyl biphenyl-2-carboxylate, tert-butyl 2-4-bromomethyl phenyl benzoate, tert-butyl 4'-bromomethyl-1,1'-biphenyl-2-carboxylate, unii-me3i28o0tc, tert-butyl 4'-bromomethyl-2-biphenylcarboxylate, me3i28o0tc, telmisartan impurity h, tert-butyl 2-4-bromomethylphenyl benzoate, 1,1-dimethylethyl 4'-bromomethyl biphenyl-2-carboxylate
PubChem CID 9906257
IUPAC Name tert-butyl 2-[4-(bromomethyl)phenyl]benzoate
SMILES CC(C)(C)OC(=O)C1=CC=CC=C1C2=CC=C(C=C2)CBr

Specifications

Quantity 1 g
Solubility Information Soluble in chloroform,ethyl acetate,and DMSO.
Formula Weight 347.24
Percent Purity 97%
Chemical Name or Material tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 442-850-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only