missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(bromomethyl)benzoate, 95%

Catalog No. p-7032920
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3554003 1 g
AAH3554009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3554003 Supplier Thermo Scientific Chemicals Supplier No. H3554003
Only null left

CAS: 108052-76-2 | C12H15BrO2 | 271.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108052-76-2
Molecular Formula C12H15BrO2
Molecular Weight (g/mol) 271.154
MDL Number MFCD04973450
InChI Key GSIBTIUXYYFCPU-UHFFFAOYSA-N
Synonym tert-butyl 4-bromomethyl benzoate, 4-bromomethylbenzoic acid tert-butyl ester, 4-bromomethyl-benzoic acid tert-butyl ester, 4-bromomethyl benzoic acid tert-butyl ester, tert-butyl p-bromomethyl benzoate, 4-bromo methyl benzoic acid mono tert-butyl ester, tert-butyl-4-bromomethylbenzoate, 1,1-dimethylethyl 4-bromomethyl benzoate, benzoic acid, 4-bromomethyl-, 1,1-dimethylethyl ester, 4-bromomethyl-benzoic acid, 1,1-dimethylethyl ester
PubChem CID 11414578
IUPAC Name tert-butyl 4-(bromomethyl)benzoate
SMILES CC(C)(C)OC(=O)C1=CC=C(C=C1)CBr

Specifications

Quantity 1 g
Formula Weight 271.15
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-(bromomethyl)benzoate
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only