missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-(5-formylpyrid-2-yl)piperazine-1-carboxylate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 479226-10-3
Molecular Formula C15H21N3O3
Molecular Weight (g/mol) 291.351
MDL Number MFCD08271923
InChI Key NETKWTAEPIGRLQ-UHFFFAOYSA-N
Synonym tert-butyl 4-5-formylpyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-5-formylpyrid-2-yl piperazine-1-carboxylate, 4-5-formylpyridin-2-yl piperazine, 4-5-formylpyridin-2-yl piperazine, n1-boc protected, 1-piperazinecarboxylicacid, 4-5-formyl-2-pyridinyl-, 1,1-dimethylethyl ester, 1-piperazinecarboxylicacid,4-5-formyl-2-pyridinyl-,1,1-dimethylethyl ester
PubChem CID 18525767
IUPAC Name tert-butyl 4-(5-formylpyridin-2-yl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC43904CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC43904DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

479226-10-3
291.351
NETKWTAEPIGRLQ-UHFFFAOYSA-N
18525767
CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)C=O
C15H21N3O3
MFCD08271923
tert-butyl 4-5-formylpyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-5-formylpyrid-2-yl piperazine-1-carboxylate, 4-5-formylpyridin-2-yl piperazine, 4-5-formylpyridin-2-yl piperazine, n1-boc protected, 1-piperazinecarboxylicacid, 4-5-formyl-2-pyridinyl-, 1,1-dimethylethyl ester, 1-piperazinecarboxylicacid,4-5-formyl-2-pyridinyl-,1,1-dimethylethyl ester
tert-butyl 4-(5-formylpyridin-2-yl)piperazine-1-carboxylate

Specifications

479226-10-3
100.0
C15H21N3O3
tert-butyl 4-5-formylpyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-5-formylpyrid-2-yl piperazine-1-carboxylate, 4-5-formylpyridin-2-yl piperazine, 4-5-formylpyridin-2-yl piperazine, n1-boc protected, 1-piperazinecarboxylicacid, 4-5-formyl-2-pyridinyl-, 1,1-dimethylethyl ester, 1-piperazinecarboxylicacid,4-5-formyl-2-pyridinyl-,1,1-dimethylethyl ester
CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)C=O
291.351
291.35
Tert-butyl 4-(5-formylpyrid-2-yl)piperazine-1-carboxylate
96.5
97%
MFCD08271923
NETKWTAEPIGRLQ-UHFFFAOYSA-N
tert-butyl 4-(5-formylpyridin-2-yl)piperazine-1-carboxylate
18525767
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents