accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 479226-10-3

CH3H3CCH3OONNNO

CAS RN 479226-10-3

IUPAC Name: tert-butyl 4-(5-formylpyridin-2-yl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(5-formylpyridin-2-yl)piperazine-1-carboxylate
Molecular Weight (g/mol): 291.35
Molecular Formula: C15H21N3O3
InChi Key: NETKWTAEPIGRLQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=C(C=O)C=C1

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.