missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC31009DA

Quantity 1g

 View more versions of this product

Catalog No. CC31009DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

864068-79-1
298.401
tert-butyl 4-4-hydroxymethyl-1,3-thiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-hydroxymethyl thiazol-2-yl piperidine-1-carboxylate, 4-4-hydroxymethyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 1-1,1-dimethylethyl 4-4-hydroxymethyl-2-thiazolyl-1-piperidine-carboxylate, 1-1,1-dimethylethyl 4-4-hydroxymethyl-2-thiazolyl-1-piperidinecarboxylate, 1-piperidinecarboxylicacid, 4-4-hydroxymethyl-2-thiazolyl-, 1,1-dimethylethyl ester
tert-butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate
C14H22N2O3S
RDZWXODJIWMJPE-UHFFFAOYSA-N
7164573
CC(C)(C)OC(=O)N1CCC(CC1)C2=NC(=CS2)CO

Specifications

tert-Butyl 4-[4-(hydroxymethyl)-1, 3-thiazol-2-yl]piperidine-1-carboxylate
95%
1g
RDZWXODJIWMJPE-UHFFFAOYSA-N
tert-butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate
7164573
95%
864068-79-1
C14H22N2O3S
tert-butyl 4-4-hydroxymethyl-1,3-thiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-hydroxymethyl thiazol-2-yl piperidine-1-carboxylate, 4-4-hydroxymethyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 1-1,1-dimethylethyl 4-4-hydroxymethyl-2-thiazolyl-1-piperidine-carboxylate, 1-1,1-dimethylethyl 4-4-hydroxymethyl-2-thiazolyl-1-piperidinecarboxylate, 1-piperidinecarboxylicacid, 4-4-hydroxymethyl-2-thiazolyl-, 1,1-dimethylethyl ester
CC(C)(C)OC(=O)N1CCC(CC1)C2=NC(=CS2)CO
298.401
298.4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents