missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate, 97%, Thermo Scientific™

Catalog No. p-4509908
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC31009CB 250mg
CC31009DA 1g
CC31009DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC31009CB Supplier Thermo Scientific Chemicals Supplier No. CC31009CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 864068-79-1
Molecular Formula C14H22N2O3S
Molecular Weight (g/mol) 298.401
InChI Key RDZWXODJIWMJPE-UHFFFAOYSA-N
Synonym tert-butyl 4-4-hydroxymethyl-1,3-thiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-hydroxymethyl thiazol-2-yl piperidine-1-carboxylate, 4-4-hydroxymethyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 1-1,1-dimethylethyl 4-4-hydroxymethyl-2-thiazolyl-1-piperidine-carboxylate, 1-1,1-dimethylethyl 4-4-hydroxymethyl-2-thiazolyl-1-piperidinecarboxylate, 1-piperidinecarboxylicacid, 4-4-hydroxymethyl-2-thiazolyl-, 1,1-dimethylethyl ester
PubChem CID 7164573
IUPAC Name tert-butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C2=NC(=CS2)CO

Specifications

Assay Percent Range 95%
Quantity 250mg
Formula Weight 298.4
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-[4-(hydroxymethyl)-1, 3-thiazol-2-yl]piperidine-1-carboxylate