missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-(4-bromobenzyloxy)piperidine-1-carboxylate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 930111-10-7
Molecular Formula C17H24BrNO3
Molecular Weight (g/mol) 370.287
MDL Number MFCD09879975
InChI Key HGSFDBSIEHXEQD-UHFFFAOYSA-N
Synonym tert-butyl 4-4-bromobenzyloxy piperidine-1-carboxylate, tert-butyl 4-4-bromobenzyl oxy piperidine-1-carboxylate, tert-butyl 4-4-bromophenyl methoxy piperidine-1-carboxylate, 4-4-bromobenzyloxy piperidine
PubChem CID 24229758
IUPAC Name tert-butyl 4-[(4-bromophenyl)methoxy]piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC67910DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC67910DE
View Documents
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

930111-10-7
370.287
HGSFDBSIEHXEQD-UHFFFAOYSA-N
24229758
CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)Br
C17H24BrNO3
MFCD09879975
tert-butyl 4-4-bromobenzyloxy piperidine-1-carboxylate, tert-butyl 4-4-bromobenzyl oxy piperidine-1-carboxylate, tert-butyl 4-4-bromophenyl methoxy piperidine-1-carboxylate, 4-4-bromobenzyloxy piperidine
tert-butyl 4-[(4-bromophenyl)methoxy]piperidine-1-carboxylate

Specifications

930111-10-7
100.0
C17H24BrNO3
tert-butyl 4-4-bromobenzyloxy piperidine-1-carboxylate, tert-butyl 4-4-bromobenzyl oxy piperidine-1-carboxylate, tert-butyl 4-4-bromophenyl methoxy piperidine-1-carboxylate, 4-4-bromobenzyloxy piperidine
CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)Br
370.287
370.28
Tert-butyl 4-(4-bromobenzyloxy)piperidine-1-carboxylate
96.5
97%
MFCD09879975
HGSFDBSIEHXEQD-UHFFFAOYSA-N
tert-butyl 4-[(4-bromophenyl)methoxy]piperidine-1-carboxylate
24229758
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents