missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-(4-bromobenzyloxy)piperidine-1-carboxylate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC67910DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC67910DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

930111-10-7
370.287
HGSFDBSIEHXEQD-UHFFFAOYSA-N
24229758
CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)Br
C17H24BrNO3
MFCD09879975
tert-butyl 4-4-bromobenzyloxy piperidine-1-carboxylate, tert-butyl 4-4-bromobenzyl oxy piperidine-1-carboxylate, tert-butyl 4-4-bromophenyl methoxy piperidine-1-carboxylate, 4-4-bromobenzyloxy piperidine
tert-butyl 4-[(4-bromophenyl)methoxy]piperidine-1-carboxylate

Specifications

Tert-butyl 4-(4-bromobenzyloxy)piperidine-1-carboxylate
96.5
97%
C17H24BrNO3
MFCD09879975
HGSFDBSIEHXEQD-UHFFFAOYSA-N
tert-butyl 4-[(4-bromophenyl)methoxy]piperidine-1-carboxylate
24229758
97%
930111-10-7
100.0
Amber Glass Bottle
5g
tert-butyl 4-4-bromobenzyloxy piperidine-1-carboxylate, tert-butyl 4-4-bromobenzyl oxy piperidine-1-carboxylate, tert-butyl 4-4-bromophenyl methoxy piperidine-1-carboxylate, 4-4-bromobenzyloxy piperidine
CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)Br
370.287
370.28
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents