missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-oxocyclobutanecarboxylate, 97%

Catalog No. AAH6206403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6206403 1 g
AAH6206406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6206403 Supplier Thermo Scientific Chemicals Supplier No. H6206403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 145549-76-4 | C9H14O3 | 170.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145549-76-4
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.208
MDL Number MFCD12827541
InChI Key JINYZTGTQXDUQR-UHFFFAOYSA-N
Synonym tert-butyl 3-oxocyclobutanecarboxylate, 3-oxo-cyclobutanecarboxylic acid tert-butyl ester, cyclobutanecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, tert-butyl 3-oxo-cyclobutanecarboxylate, 1-boc-cyclobutan-3-one, t-butyl 3-oxocyclobutanecarboxylate, 3-tert-butyloxycarbonylcyclobutanone, tert-butyl3-oxocyclobutanecarboxylate, tert-butyl 3-oxocyclobutane carboxylate, t-butyl-3-oxocyclobutanecarboxylate
PubChem CID 10261520
IUPAC Name tert-butyl 3-oxocyclobutane-1-carboxylate
SMILES CC(C)(C)OC(=O)C1CC(=O)C1

Specifications

Quantity 1 g
Formula Weight 170.2
Percent Purity 97%
Chemical Name or Material tert-Butyl 3-oxocyclobutanecarboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only