CAS RN 145549-76-4
IUPAC Name:
tert-butyl 3-oxocyclobutane-1-carboxylate
Synonyms:
tert-butyl 3-oxocyclobutane-1-carboxylate
Molecular Weight (g/mol):
170.21
Molecular Formula:
C9H14O3
InChi Key:
JINYZTGTQXDUQR-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)C1CC(=O)C1
1
–
1
of
1
results
1
Promotions Available
tert-Butyl 3-oxocyclobutanecarboxylate, 97%
tert-Butyl 3-oxocyclobutanecarboxylate, 97%
CAS: 145549-76-4 Molecular Formula: C9H14O3 Molecular Weight (g/mol): 170.208 MDL Number: MFCD12827541 InChI Key: JINYZTGTQXDUQR-UHFFFAOYSA-N Synonym: tert-butyl 3-oxocyclobutanecarboxylate,3-oxo-cyclobutanecarboxylic acid tert-butyl ester,cyclobutanecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester,tert-butyl 3-oxo-cyclobutanecarboxylate,1-boc-cyclobutan-3-one,t-butyl 3-oxocyclobutanecarboxylate,3-tert-butyloxycarbonylcyclobutanone,tert-butyl3-oxocyclobutanecarboxylate,tert-butyl 3-oxocyclobutane carboxylate,t-butyl-3-oxocyclobutanecarboxylate PubChem CID: 10261520 IUPAC Name: tert-butyl 3-oxocyclobutane-1-carboxylate SMILES: CC(C)(C)OC(=O)C1CC(=O)C1