missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 3-(Isopropyl)carbazate 98.0+%, TCI America™
Supplier: TCI America B43751G
Specifications
| tert-Butyl 3-(Isopropyl)carbazate | |
| 54°C | |
| 80°C | |
| MFCD14635748 | |
| 3-(Isopropyl)carbazic Acid tert-Butyl Ester, 1-tert-Butoxycarbonyl-2-isopropylhydrazine, 1-Boc-2-isopropylhydrazine | |
| CC(C)NNC(=O)OC(C)(C)C | |
| 174.24 | |
| 174.24 | |
| Crystalline Powder |
| 16689-35-3 | |
| White | |
| C8H18N2O2 | |
| 1 g | |
| PUAKAEDMCXRDPR-UHFFFAOYSA-N | |
| N'-(propan-2-yl)(tert-butoxy)carbohydrazide | |
| 11217534 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 16689-35-3 | |
| 174.24 | |
| PUAKAEDMCXRDPR-UHFFFAOYSA-N | |
| 11217534 | |
| CC(C)NNC(=O)OC(C)(C)C |
| C8H18N2O2 | |
| MFCD14635748 | |
| 3-(Isopropyl)carbazic Acid tert-Butyl Ester, 1-tert-Butoxycarbonyl-2-isopropylhydrazine, 1-Boc-2-isopropylhydrazine | |
| N'-(propan-2-yl)(tert-butoxy)carbohydrazide |
Safety and Handling
TSCA : No