accessibility menu, dialog, popup

UserName

CAS RN 16689-35-3

IUPAC Name: N'-(propan-2-yl)(tert-butoxy)carbohydrazide
Synonyms: N'-isopropyltert-butoxycarbohydrazide
Molecular Weight (g/mol): 174.24
Molecular Formula: C8H18N2O2
InChi Key: PUAKAEDMCXRDPR-UHFFFAOYSA-N
SMILES: CC(C)NNC(=O)OC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results