missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-Butenoate 95.0+%, TCI America™

Catalog No. p-7123181
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B38115G 5 g
B381125G 25 g
2 options

Catalog No. B38115G

Supplier: TCI America B38115G

Only null left

Chemical Identifiers

CAS 14036-55-6
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD06411714
InChI Key NGASWKRTXGWPNN-UHFFFAOYSA-N
Synonym tert-butyl 3-butenoate, tert-butyl vinylacetate, 3-butenoic acid tert-butyl ester, 3-butenoic acid, 1,1-dimethylethyl ester, t-butyl vinylacetate, tert-butyl3-butenoate, tert-butyl-3-butenoate, acmc-20ajl7, vinylacetic acid tert-butyl ester, but-3-enoic acid tert-butyl ester
PubChem CID 639778
IUPAC Name tert-butyl but-3-enoate
SMILES CC(C)(C)OC(=O)CC=C

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 g
UN Number 3272
Formula Weight 142.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl 3-Butenoate