missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 2-Bromopropionate 97.0+%, TCI America™
$115.51 - $1151.25
Chemical Identifiers
| CAS | 39149-80-9 |
|---|---|
| Molecular Formula | C7H13BrO2 |
| Molecular Weight (g/mol) | 209.083 |
| MDL Number | MFCD00043897 |
| InChI Key | CVAWKJKISIPBOD-UHFFFAOYSA-N |
| Synonym | 2-Bromopropionic Acid tert-Butyl Ester |
| PubChem CID | 545327 |
| IUPAC Name | tert-butyl 2-bromopropanoate |
| SMILES | CC(C(=O)OC(C)(C)C)Br |
Chemical Identifiers
| 39149-80-9 | |
| 209.083 | |
| CVAWKJKISIPBOD-UHFFFAOYSA-N | |
| 545327 | |
| CC(C(=O)OC(C)(C)C)Br |
| C7H13BrO2 | |
| MFCD00043897 | |
| 2-Bromopropionic Acid tert-Butyl Ester | |
| tert-butyl 2-bromopropanoate |
Specifications
| 39149-80-9 | |
| 62°C | |
| MFCD00043897 | |
| 2-Bromopropionic Acid tert-Butyl Ester | |
| CC(C(=O)OC(C)(C)C)Br | |
| 209.083 | |
| 209.08 | |
| Liquid |
| Yellow | |
| C7H13BrO2 | |
| 25 g | |
| CVAWKJKISIPBOD-UHFFFAOYSA-N | |
| tert-butyl 2-bromopropanoate | |
| 545327 | |
| ≥97.0% (GC) | |
| tert-Butyl 2-Bromopropionate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator