missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate 98.0+%, TCI America™
$54.06 - $160.30
Chemical Identifiers
| CAS | 125995-13-3 |
|---|---|
| Molecular Formula | C14H28NO4 |
| Molecular Weight (g/mol) | 274.38 |
| MDL Number | MFCD07369252 |
| InChI Key | HWSHVKNLMBMKSR-GHMZBOCLSA-O |
| Synonym | 4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, tert-butyl 2-4r,6r-6-2-aminoethyl-2,2-dimethyl-1,3-dioxan-4-yl acetate, 4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,cis-1,1-dimethylethyl-6-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3, 4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4acetate, +/--cis-1,1-dimethylethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r-2,2-dimethyl-6beta-2-aminoethyl-1,3-dioxane-4beta-acetic acid tert-butyl ester, 4r-cis-1,1-dimethyethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate |
| PubChem CID | 2734288 |
| IUPAC Name | 2-[(4R,6R)-6-[2-(tert-butoxy)-2-oxoethyl]-2,2-dimethyl-1,3-dioxan-4-yl]ethan-1-aminium |
| SMILES | CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC[NH3+])OC(C)(C)O1 |
Chemical Identifiers
| 125995-13-3 | |
| 274.38 | |
| HWSHVKNLMBMKSR-GHMZBOCLSA-O | |
| 2734288 | |
| CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC[NH3+])OC(C)(C)O1 |
| C14H28NO4 | |
| MFCD07369252 | |
| 4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, tert-butyl 2-4r,6r-6-2-aminoethyl-2,2-dimethyl-1,3-dioxan-4-yl acetate, 4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,cis-1,1-dimethylethyl-6-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3, 4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4acetate, +/--cis-1,1-dimethylethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r-2,2-dimethyl-6beta-2-aminoethyl-1,3-dioxane-4beta-acetic acid tert-butyl ester, 4r-cis-1,1-dimethyethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate | |
| 2-[(4R,6R)-6-[2-(tert-butoxy)-2-oxoethyl]-2,2-dimethyl-1,3-dioxan-4-yl]ethan-1-aminium |
Specifications
| 125995-13-3 | |
| C14H28NO4 | |
| 1 g | |
| HWSHVKNLMBMKSR-GHMZBOCLSA-O | |
| 2-[(4R,6R)-6-[2-(tert-butoxy)-2-oxoethyl]-2,2-dimethyl-1,3-dioxan-4-yl]ethan-1-aminium | |
| 2734288 | |
| ≥98.0% (GC) | |
| tert-Butyl 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate |
| Yellow | |
| MFCD07369252 | |
| 4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, tert-butyl 2-4r,6r-6-2-aminoethyl-2,2-dimethyl-1,3-dioxan-4-yl acetate, 4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,cis-1,1-dimethylethyl-6-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3, 4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4acetate, +/--cis-1,1-dimethylethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r-2,2-dimethyl-6beta-2-aminoethyl-1,3-dioxane-4beta-acetic acid tert-butyl ester, 4r-cis-1,1-dimethyethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate | |
| CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC[NH3+])OC(C)(C)O1 | |
| 274.38 | |
| 273.37 | |
| Liquid |
Safety and Handling
TSCA : No