missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate 98.0+%, TCI America™

Catalog No. B37021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37021G 1 g
B37025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37021G Supplier TCI America Supplier No. B37021G

Chemical Identifiers

CAS 125995-13-3
Molecular Formula C14H28NO4
Molecular Weight (g/mol) 274.38
MDL Number MFCD07369252
InChI Key HWSHVKNLMBMKSR-GHMZBOCLSA-O
Synonym 4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, tert-butyl 2-4r,6r-6-2-aminoethyl-2,2-dimethyl-1,3-dioxan-4-yl acetate, 4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,cis-1,1-dimethylethyl-6-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3, 4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4acetate, +/--cis-1,1-dimethylethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r-2,2-dimethyl-6beta-2-aminoethyl-1,3-dioxane-4beta-acetic acid tert-butyl ester, 4r-cis-1,1-dimethyethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate
PubChem CID 2734288
IUPAC Name 2-[(4R,6R)-6-[2-(tert-butoxy)-2-oxoethyl]-2,2-dimethyl-1,3-dioxan-4-yl]ethan-1-aminium
SMILES CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC[NH3+])OC(C)(C)O1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 273.37
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate