missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2,2,2-trichloroacetimidate, 95%, Thermo Scientific™

Chemical Identifiers

CAS 98946-18-0
Molecular Formula C6H10Cl3NO
Molecular Weight (g/mol) 218.50
MDL Number MFCD00077410
InChI Key CQXDYHPBXDZWBA-UHFFFAOYSA-N
Synonym tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine
PubChem CID 2734700
IUPAC Name tert-butyl 2,2,2-trichloroethanimidate
SMILES CC(C)(C)OC(=N)C(Cl)(Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC369330050
SDS View Documents
Thermo Scientific Chemicals
369330050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC369330250
SDS View Documents
Thermo Scientific Chemicals
369330250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC369331000
SDS View Documents
Thermo Scientific Chemicals
369331000
100g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Alkylation reagent
Specifications

Chemical Identifiers

98946-18-0
218.50
CQXDYHPBXDZWBA-UHFFFAOYSA-N
2734700
CC(C)(C)OC(=N)C(Cl)(Cl)Cl
C6H10Cl3NO
MFCD00077410
tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine
tert-butyl 2,2,2-trichloroethanimidate

Specifications

98946-18-0
64.0°C to 66.0°C (15.0mmHg)
Authentic
Glass bottle
1.4550 to 1.4570
MFCD00077410
1.222
CQXDYHPBXDZWBA-UHFFFAOYSA-N
tert-butyl 2,2,2-trichloroethanimidate
2734700
95%
1.2220g/mL
55°C
94% min. (GC)
C6H10Cl3NO
CCl3C(=NH)OC(CH3)3
5g
tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine
CC(C)(C)OC(=N)C(Cl)(Cl)Cl
218.50
218.51
tert-Butyl 2, 2, 2-trichloroacetimidate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only