missing translation for 'onlineSavingsMsg'
Learn More

tert-Butoxybis(dimethylamino)methane, tech. 90%

Catalog No. AAH2786103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2786103 1 g
AAH2786106 5 g
AAH2786114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2786103 Supplier Thermo Scientific Chemicals Supplier No. H2786103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5815-08-7 | C9H24N2O | 176.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5815-08-7
Molecular Formula C9H24N2O
Molecular Weight (g/mol) 176.30
MDL Number MFCD00042858
InChI Key HXRAMSFGUAOAJR-UHFFFAOYSA-P
Synonym tert-butoxy bis dimethylamino methane, 1-tert-butoxy-n,n,n',n'-tetramethylmethanediamine, tert-butoxybis dimethylamino methane, bredereck's reagent, bredereck reagent, tert-butoxy dimethylamino methyl dimethylamine, unii-jej0u558f7, bis dimethylamino-t-butoxymethane, tert-butoxy-bis dimethylamino methane, bis dimethylamino tert-butoxy methane
PubChem CID 79885
IUPAC Name N,N,N',N'-tetramethyl-1-[(2-methylpropan-2-yl)oxy]methanediamine
SMILES CC(C)(C)OC(N(C)C)N(C)C

Specifications

Density 0.844
Boiling Point 50°C to 55°C (15 mmHg)
Flash Point 41°C (106°F)
Refractive Index 1.422
Linear Formula (CH3)3COCH[N(CH3)2]2
Quantity 1 g
UN Number UN1993
Formula Weight 174.3
Percent Purity 90%
Grade Technical
Chemical Name or Material tert-Butoxybis(dimethylamino)methane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 227-383-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only