accessibility menu, dialog, popup

UserName

H3CNHCH3OCH3CH3H3CNHCH3H3C

CAS RN 5815-08-7

IUPAC Name: [(tert-butoxy)(dimethylazaniumyl)methyl]dimethylazanium
Synonyms: [tert-butoxy(dimethylammonio)methyl]dimethylazanium
Molecular Weight (g/mol): 176.3
Molecular Formula: C9H24N2O
InChi Key: HXRAMSFGUAOAJR-UHFFFAOYSA-P
SMILES: C[NH+](C)C(OC(C)(C)C)[NH+](C)C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)