missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Amylamine 98.0+%, TCI America™
Supplier: TCI America A1002500ML
Specifications
| tert-Amylamine | |
| Yellow | |
| C5H13N | |
| 500 mL | |
| tert-amylamine, tert-pentylamine, 2-butanamine, 2-methyl, t-amylamine, 1,1-dimethylpropylamine, 2-methyl-2-butylamine, 2-amino-2-methylbutane, 2-methyl-butan-2-amine, 1,1-dimethyl-1-propylamine, t-pentylamine | |
| CCC(C)(C)N | |
| 87.166 | |
| CHEBI:84236 | |
| ≥98.0% (GC,T) | |
| Liquid |
| 594-39-8 | |
| 77°C | |
| MFCD00008056 | |
| 1106 | |
| GELMWIVBBPAMIO-UHFFFAOYSA-N | |
| 2-methylbutan-2-amine | |
| 68986 | |
| 87.17 | |
| Reagent |
Chemical Identifiers
| 594-39-8 | |
| 87.166 | |
| GELMWIVBBPAMIO-UHFFFAOYSA-N | |
| 68986 | |
| 2-methylbutan-2-amine |
| C5H13N | |
| MFCD00008056 | |
| tert-amylamine, tert-pentylamine, 2-butanamine, 2-methyl, t-amylamine, 1,1-dimethylpropylamine, 2-methyl-2-butylamine, 2-amino-2-methylbutane, 2-methyl-butan-2-amine, 1,1-dimethyl-1-propylamine, t-pentylamine | |
| CHEBI:84236 | |
| CCC(C)(C)N |
Safety and Handling
EINECSNumber : (2)-0133
TSCA : Yes