missing translation for 'onlineSavingsMsg'
Learn More

tert-Amylamine 98.0+%, TCI America™

Catalog No. p-7117181
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
A100225ML 25 mL
A1002500ML 500 mL
2 options

Catalog No. A100225ML

Supplier: TCI America A100225ML

Only null left

Chemical Identifiers

CAS 594-39-8
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008056
InChI Key GELMWIVBBPAMIO-UHFFFAOYSA-N
Synonym tert-amylamine, tert-pentylamine, 2-butanamine, 2-methyl, t-amylamine, 1,1-dimethylpropylamine, 2-methyl-2-butylamine, 2-amino-2-methylbutane, 2-methyl-butan-2-amine, 1,1-dimethyl-1-propylamine, t-pentylamine
PubChem CID 68986
ChEBI CHEBI:84236
IUPAC Name 2-methylbutan-2-amine
SMILES CCC(C)(C)N

Specifications

Color Yellow
Boiling Point 77°C
Quantity 25 mL
UN Number 1106
Formula Weight 87.17
Percent Purity ≥98.0% (GC,T)
Grade Reagent
Physical Form Liquid
Chemical Name or Material tert-Amylamine
EINECSNumber (2)-0133
TSCA Yes