missing translation for 'onlineSavingsMsg'
Learn More

tert-Amyl Ethyl Ether 98.0+%, TCI America™

Catalog No. p-7117573
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16735G 5 g
A167325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16735G Supplier TCI America Supplier No. A16735G
Only null left

Chemical Identifiers

CAS 919-94-8
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD01727266
InChI Key KFRVYYGHSPLXSZ-UHFFFAOYSA-N
Synonym 1,1-Dimethylpropyl Ethyl Ether, Ethyl tert-Pentyl Ether
PubChem CID 13527
IUPAC Name 2-ethoxy-2-methylbutane
SMILES CCC(C)(C)OCC

Specifications

Color Colorless
Boiling Point 102°C
Quantity 5 g
UN Number 3271
Formula Weight 116.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Amyl Ethyl Ether
RTECSNumber KO0351000
TSCA Yes