accessibility menu, dialog, popup

UserName

CAS RN 919-94-8

CH3OCH3H3CH3C

CAS RN 919-94-8

IUPAC Name: 2-ethoxy-2-methylbutane
Synonyms: ethyl(tert-amyl) ether
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: KFRVYYGHSPLXSZ-UHFFFAOYSA-N
SMILES: CCOC(C)(C)CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results