missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Amyl Ethyl Ether 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 102°C |
| Quantity | 5 g |
| UN Number | 3271 |
| Formula Weight | 116.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | tert-Amyl Ethyl Ether |
Safety and Handling
| RTECSNumber | KO0351000 |
| TSCA | Yes |