missing translation for 'onlineSavingsMsg'
Learn More

alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol

Catalog No. p-194715
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC203740500 50 g Glass bottle
AC203742500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC203740500 Supplier Thermo Scientific Chemicals Supplier No. 203740500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-55-5 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-55-5 , 586-81-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001557
InChI Key WUOACPNHFRMFPN-UHFFFAOYSA-N
Synonym alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
PubChem CID 17100
ChEBI CHEBI:22469
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
SMILES CC1=CCC(CC1)C(C)(C)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 18°C
Color Colorless to Yellow
Density 0.9330g/mL
Boiling Point 214°C to 224°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range gamma-terpineol: balance (GC)
Packaging Glass bottle
Refractive Index 1.4820 to 1.485
Quantity 50 g
Beilstein 06, II, 66
Specific Gravity 0.933
Merck Index 15, 9313
Solubility Information Solubility in water: negligible. Other solubilities: soluble in organic solvents ,diethyl ether,,ethanol,benzene,propylene glycol
Formula Weight 154.25
Percent Purity 97%
Concentration or Composition (by Analyte or Components) alpha-terpineol: 85 to 90%, gamma-terpineol: balance (GC)
Physical Form Liquid
Chemical Name or Material α-Terpineol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-680-6
TSCA TSCA

RUO – Research Use Only