missing translation for 'onlineSavingsMsg'
Learn More

γ-Terpinene, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019106
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019106 1 mL
111018254 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019106 Supplier Merck Emd Millipore Supplier No. 864761ML

γ-Terpinene is a monoterpene hydrocarbon present in essential oils extracted from Thymus vulgaris L., and exhibits antimicrobial, carminative and expectorant activities.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 99-85-4
Vapor Pressure ˜0.7 mmHg (20°C)
Vapor Density 4.7 (vs. air)
Density 0.85 g/mL (at 25°C)
Flash Point 50°C - closed cup
Physical Form Neat
Linear Formula C10H16
MDL Number MFCD00001537
Boiling Point 182°C
Synonym 1-Isopropyl-4-methyl-1,4-cyclohexadiene; p-Mentha-1,4-diene
Type Analytical Standard
Quantity 1 mL
Formula Weight 136.23
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less