missing translation for 'onlineSavingsMsg'
Learn More

γ-Terpinene, ≥98.5% (GC), MilliporeSigma™ Supelco™

γ-Terpinene is a monoterpene hydrocarbon present in essential oils extracted from Thymus vulgaris L., and exhibits antimicrobial, carminative and expectorant activities.

Supplier:  Merck Emd Millipore 864765ML

 View more versions of this product

Catalog No. 111018254

Only null left

Description

Description

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

99-85-4
4.7 (vs. air)
50°C - closed cup
C10H16
182°C
Analytical Standard
136.23
Analytical Standard
˜0.7 mmHg (20°C)
0.85 g/mL (at 25°C)
Neat
MFCD00001537
1-Isopropyl-4-methyl-1,4-cyclohexadiene; p-Mentha-1,4-diene
5 mL
≥98.5% (GC)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions