missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Teriflunomide

Chemical Identifiers

CAS 163451-81-8
Chemical Name or Material Teriflunomide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC467110010
View Documents
Thermo Scientific™
467110010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC467112500
View Documents
Thermo Scientific™
467112500
250 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

163451-81-8
Teriflunomide

Specifications

163451-81-8
1 g
229.0°C
Teriflunomide
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only