missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tergitol(R) TMN-6, Surfactant, Spectrum™ Chemical
T1303, 60828-78-6, C12H25O(CH2CH2O)nH
$950.29
Specifications
| Boiling Point | 106°C (223°F) |
|---|---|
| Composition | C12H26O(CH2CH2O)n (for Polyethylene glycol timethylnonyl ether) |
Description
Spectrum™ Chemical Tergitol(R) TMN-6, Surfactant is a is a water-soluble, nonionic surfactant that is a superior wetting agent for use in multiple applications including paper & textile processing and hard surface cleaners.Specifications
| 106°C (223°F) | |
| C12H26O(CH2CH2O)n (for Polyethylene glycol timethylnonyl ether) |