Learn More
Tergitol(R) TMN-6, Surfactant, Spectrum™ Chemical
T1303, 60828-78-6, C12H25O(CH2CH2O)nH
Supplier:  Spectrum Chemical Mfg Cor T13032.5LTGL
 View more versions of this product
Catalog No. 18607293
Only null left
Description
Description
Spectrum™ Chemical Tergitol(R) TMN-6, Surfactant is a is a water-soluble, nonionic surfactant that is a superior wetting agent for use in multiple applications including paper & textile processing and hard surface cleaners.
Specifications
Chemical Identifiers
106°C (223°F)
C12H26O(CH2CH2O)n (for Polyethylene glycol timethylnonyl ether)
Specifications
106°C (223°F)
C12H26O(CH2CH2O)n (for Polyethylene glycol timethylnonyl ether)
2.5 L
Product Suggestions
Product Suggestions